Anti-BSX Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-12905R-A555
BOSSBS-12905R-A555EA 893 EUR
BOSSBS-12905R-A555
Anti-BSX Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
BSX(Brain-specific homeobox protein homolog) may play a role in the determination and function of cell types in the brain. In brain, it is restricted to a few specific developing brain structures such as pineal gland, telencephalic septum, hypothalamic pre-mammillary body and arcuate nucleus. Bsx might be considered an important molecular marker for early embryonic stages of epiphysis development. It belongs to the distal-less homeobox family and contains 1 homeobox DNA-binding domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BSX
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR