Anti-ODC Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-1294R-A647
BOSSBS-1294R-A647EA 901 EUR
BOSSBS-1294R-A647
Anti-ODC Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Transports C5-C7 oxodicarboxylates across the inner membranes of mitochondria. Can transport 2-oxoadipate, 2-oxoglutarate, adipate, glutarate, and to a lesser extent, pimelate, 2-oxopimelate, 2-aminoadipate, oxaloacetate, and citrate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ODC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR