Anti-ODC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1294R-A680
BOSSBS-1294R-A680EA 899 EUR
BOSSBS-1294R-A680
Anti-ODC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Transports C5-C7 oxodicarboxylates across the inner membranes of mitochondria. Can transport 2-oxoadipate, 2-oxoglutarate, adipate, glutarate, and to a lesser extent, pimelate, 2-oxopimelate, 2-aminoadipate, oxaloacetate, and citrate.

Type: Primary
Antigen: ODC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 321-461/461
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR