Anti-ACSF4 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-12951R-CY7
BOSSBS-12951R-CY7EA 1350 EUR
BOSSBS-12951R-CY7
Anti-ACSF4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
AASDH is an Acyl CoA synthetase. Acyl CoA synthases catalyze the initial reaction in fatty acid metabolism, by forming a thioester with CoA .

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACSF4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR