Anti-Acetyl Coenzyme A Carboxylase beta Ser220 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12952R-A680
BOSSBS-12952R-A680EA 917 EUR
BOSSBS-12952R-A680
Anti-Acetyl Coenzyme A Carboxylase beta Ser220 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyses the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene.

Type: Primary
Antigen: Acetyl Coenzyme A Carboxylase beta Ser220
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR