Anti-Acetyl Coenzyme A Carboxylase alpha Ser80 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Lieferant: Bioss
BS-12954R-A750
BOSSBS-12954R-A750EA 917 EUR
BOSSBS-12954R-A750
Anti-Acetyl Coenzyme A Carboxylase alpha Ser80 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyses the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene.
Type: Primary
Antigen: Acetyl Coenzyme A Carboxylase alpha Ser80
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen