Anti-DGKE Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-12999R-CY3
BOSSBS-12999R-CY3EA 900 EUR
BOSSBS-12999R-CY3
Anti-DGKE Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Highly selective for arachidonate-containing species of diacylglycerol (DAG). May terminate signals transmitted through arachidonoyl-DAG or may contribute to the synthesis of phospholipids with defined fatty acid composition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DGKE
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR