Anti-DHRS7B Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13000R
BOSSBS-13000REA 1090 EUR
BOSSBS-13000R
Anti-DHRS7B Rabbit Polyclonal Antibody
Antikörper
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function. [provided by RefSeq, Jul 2008].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DHRS7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR