Anti-Dishevelled 2 Ser143 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13004R-A750
BOSSBS-13004R-A750EA 917 EUR
BOSSBS-13004R-A750
Anti-Dishevelled 2 Ser143 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Participates in Wnt signaling by binding to the cytoplasmic C-terminus of frizzled family members and transducing the Wnt signal to down-stream effectors. Promotes internalization and degradation of frizzled proteins upon Wnt signaling. Plays a role both in canonical and non-canonical Wnt signaling. Plays a role in the signal transduction pathways mediated by multiple Wnt genes (By similarity).

Type: Primary
Antigen: Dishevelled 2 Ser143
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR