Anti-DTYMK Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13034R
BOSSBS-13034REA 725 EUR
BOSSBS-13034R
Anti-DTYMK Rabbit Polyclonal Antibody
Antikörper
DTYMK is a 212 amino acid protein that belongs to the thymidylate kinase family and is involved in pyrimidine metabolism. Specifically, DTYMK catalyzes the ATP-dependent conversion of dTMP (deoxythymidine monophosphate) to dTDP (deoxythymidine diphosphate), which then functions as one of the four nucleotides in DNA. Via its role in the catalytic creation of dTDP, DTYMK plays an important role in the pathway of DNA synthesis and is thought to be involved in cell cycle progression and cell growth. DTYMK expression levels peak during the S phase (synthesis phase) of the cell cycle, further supporting the role of DTYMK in DNA synthesis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DTYMK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR