Anti-DTYMK/CDC8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-13034R-A750
BOSSBS-13034R-A750EA 890 EUR
BOSSBS-13034R-A750
Anti-DTYMK/CDC8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
DTYMK is a 212 amino acid protein that belongs to the thymidylate kinase family and is involved in pyrimidine metabolism. Specifically, DTYMK catalyzes the ATP-dependent conversion of dTMP (deoxythymidine monophosphate) to dTDP (deoxythymidine diphosphate), which then functions as one of the four nucleotides in DNA. Via its role in the catalytic creation of dTDP, DTYMK plays an important role in the pathway of DNA synthesis and is thought to be involved in cell cycle progression and cell growth. DTYMK expression levels peak during the S phase (synthesis phase) of the cell cycle, further supporting the role of DTYMK in DNA synthesis.
Type: Primary
Antigen: DTYMK/CDC8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/212
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen