Anti-DTYMK Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-13034R-CY3
BOSSBS-13034R-CY3EA 900 EUR
BOSSBS-13034R-CY3
Anti-DTYMK Rabbit Polyclonal Antibody (Cy3®)
Antikörper
DTYMK is a 212 amino acid protein that belongs to the thymidylate kinase family and is involved in pyrimidine metabolism. Specifically, DTYMK catalyzes the ATP-dependent conversion of dTMP (deoxythymidine monophosphate) to dTDP (deoxythymidine diphosphate), which then functions as one of the four nucleotides in DNA. Via its role in the catalytic creation of dTDP, DTYMK plays an important role in the pathway of DNA synthesis and is thought to be involved in cell cycle progression and cell growth. DTYMK expression levels peak during the S phase (synthesis phase) of the cell cycle, further supporting the role of DTYMK in DNA synthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DTYMK
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR