Anti-DYX1C1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-13043R-CY7
BOSSBS-13043R-CY7EA 900 EUR
BOSSBS-13043R-CY7
Anti-DYX1C1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The gene encoding DYX1C1 maps in the 15q21 region, which is disrupted by a translocation t(2;15)(q11;q21) and segregates with dyslexia. Two sequence changes in DYX1C1, including one involving the translation initiation sequence and an Elk-1 transcription factor binding site (-3G -->A) and a codon (1249G -->T), introduce a premature stop codon and truncate the protein by 4 amino acids. DYX1C1 encodes a nuclear tetratricopeptide repeat domain protein that is dynamically regulated in brain. In human brain, DYX1C1 protein localizes to a fraction of cortical neurons and white matter glial cells. It is also expressed in lung, kidney and testis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DYX1C1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR