Anti-ECH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-13048R-A488
BOSSBS-13048R-A488EA 893 EUR
BOSSBS-13048R-A488
Anti-ECH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ECH1 is a 328 amino acid protein that localizes to both the mitochondrion and the peroxisome and belongs to the hydratase/isomerase superfamily. Existing as a homohexamer, ECH1 is involved in the fatty acid-beta oxidation pathway, specifically functioning to catalyze the isomerization of 3-trans,5-cis-dienoyl-CoA to 2-trans,4-trans-dienoyl-CoA. The gene encoding ECH1 maps to human chromosome 19, which is the genetic home for a number of immunoglobulin superfamily members, including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG family and Fc receptors (FcRs).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ECH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR