Anti-FABP4 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-13063R
BOSSBS-13063REA 743 EUR
BOSSBS-13063R
Anti-FABP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat