Anti-eNOS Ser633 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13074R-A750
BOSSBS-13074R-A750EA 895 EUR
BOSSBS-13074R-A750
Anti-eNOS Ser633 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
eNOS is a calcium/calmodulin dependent enzyme which undergoes several post translational modifications, including acylation with myristate and palmitate, and phosphorylation on numerous residues. As with the other members of the NOS family, eNOS derives the diffusible multifunctional second messenger NO from L arginine through a series of reactions in which L citrulline is an intermediate. eNOS plays an important role in controlling vascular tone, platelet aggregation, and cardiac myocyte function.

Type: Primary
Antigen: eNOS Ser633
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR