Anti-SLC29A2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13078R
BOSSBS-13078REA 1090 EUR
BOSSBS-13078R
Anti-SLC29A2 Rabbit Polyclonal Antibody
Antikörper
Equilibrative nucleoside transporters (ENTs) regulate many physiological processes and are widely distributed in mammals, plants, yeasts, insects, nematodes and protozoans. They enable facilitated diffusion of hydrophilic nucleosides, such as adenosine and nucleoside analogs, across cell membranes. ENTs are required for uptake of antiviral and anticancer nucleoside drugs and influence a variety of physiological processes, such as neurotransmission and platelet aggregation, by regulating the amount of adenoside available to cell surface receptors. Equilibrative nucleoside transporter 2 (ENT2), also designated solute carrier family 29 (nucleoside transporters), member 2, belongs to the SLC29A transporter family and is a mammalian ENT isoform. ENT2 mediates the equilibrative transport of hypoxanthine in addition to nucleosides and is purine-selective.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SLC29A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR