Anti-ETFDH Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13110R-A680
BOSSBS-13110R-A680EA 890 EUR
BOSSBS-13110R-A680
Anti-ETFDH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.

Type: Primary
Antigen: ETFDH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-150/617
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR