Anti-EYA4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-13126R-A555
BOSSBS-13126R-A555EA 893 EUR
BOSSBS-13126R-A555
Anti-EYA4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
A gene of chromosome 6q23 encodes the 640 amino acid protein, EYA4 (eyes absent) (1). EYA is one of four members of the eyes absent family (1). A 271 amino acid domain at the carboxyl terminal is highly conserved amongst the members of the eyes absent family (1). EYA4 is expressed in the craniofacial mesenchyme, the dermamyotome, and the limb (1). The conserved region in other EYA proteins interacts with SIX, DACH, and G-proteins, which regulate transcription in early embryonic development (1,2,3,4). SIX translocates EYA1-3 to the nucleus, and G-proteins can stop this interaction (3,4).More Product Information