Anti-IGF2BP3 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1312R-CY3
BOSSBS-1312R-CY3EA 900 EUR
BOSSBS-1312R-CY3
Anti-IGF2BP3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
RNA-binding factor that may recruits target transcripts to cytoplasmic protein-RNA complexes (mRNPs). This transcript 'caging' into mRNPs allows mRNA transport and transient storage. It also modulates the rate and location at which target transcripts encounter the translational apparatus and shields them from endonuclease attacks or microRNA-mediated degradation. Binds to the 3'-UTR of CD44 mRNA and stabilizes it, hence promotes cell adhesion and invadopodia formation in cancer cells. Binds to beta-actin/ACTB and MYC transcripts. Binds to the 5'-UTR of the insulin-like growth factor 2 (IGF2)More Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol IGF2BP3
Antigen-Name Insulin-like growth Factor 2 mRNA binding Protein 3
Antigen-Synonyme IMP-3|KOC|IMP3|CT98|VICKZ3|KOC1
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy3®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 10643
Isotyp IgG
ImmunoChemie Yes
Immunogen 110-170/579
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR