Anti-FANCD2 Ser1404 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13141R-A680
BOSSBS-13141R-A680EA 917 EUR
BOSSBS-13141R-A680
Anti-FANCD2 Ser1404 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Fanconi Anemia (FANC) is a human autosomal-recessive cancer susceptibility disorder characterized by congenital defects, progressive bone marrow failure, and cellular hypersensitivity to mitomycin C (MMC). The FANC subunit D2 protein is vital for cellular resistance to DNA cross-linking and the arrest of DNA synthesis after ionizing radiation. DNA damage activates the monoubiquitination of FANC D2, targeting nuclear foci containing the BRCA 1 protein.

Type: Primary
Antigen: FANCD2 Ser1404
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR