Anti-Ferredoxin Reductase/ADXR Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13154R-A750
BOSSBS-13154R-A750EA 890 EUR
BOSSBS-13154R-A750
Anti-Ferredoxin Reductase/ADXR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Adrenodoxin Reductase (ADX Reductase) is a mitochondrial flavoprotein that receives electrons from NADPH and thereby initiates the electron-transport chain serving mitochondrial cytochromes P450. ADX Reductase participates in cholesterol side chain cleavage in all steroidogenic tissues, steroid 11-? hydroxylation in the adrenal cortex, 25-OH-vitamin D3-24 hydroxylation in the kidney and sterol C-27 hydroxylation in the liver. Alternate splicing of ADX Reductase produces two isoforms. Human ADX Reductase maps to human chromosome 17q24-q25.

Type: Primary
Antigen: Ferredoxin Reductase/ADXR
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-491/491
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR