Anti-FDXR Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-13154R-CY5
BOSSBS-13154R-CY5EA 893 EUR
BOSSBS-13154R-CY5
Anti-FDXR Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Adrenodoxin Reductase (ADX Reductase) is a mitochondrial flavoprotein that receives electrons from NADPH and thereby initiates the electron-transport chain serving mitochondrial cytochromes P450. ADX Reductase participates in cholesterol side chain cleavage in all steroidogenic tissues, steroid 11-? hydroxylation in the adrenal cortex, 25-OH-vitamin D3-24 hydroxylation in the kidney and sterol C-27 hydroxylation in the liver. Alternate splicing of ADX Reductase produces two isoforms. Human ADX Reductase maps to human chromosome 17q24-q25.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Ferredoxin Reductase/ADXR
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR