Anti-FIP1L1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-13173R-A555
BOSSBS-13173R-A555EA 893 EUR
BOSSBS-13173R-A555
Anti-FIP1L1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The Component of the Cleavage and Polyadenylation Specificity Factor (CPSF) complex plays an important role in the 3'-end formation of pre-mRNA. This complex recognizes the AAUAAA signal sequence and interacts with poly(A) polymerase to process and add to the poly(A) tail. FIP1L1 (FIP1-like 1), also known as Pre-mRNA 3'-end-processing factor FIP1, FIP1 (Factor interacting with PAP) and RHE (Rearranged in hypereosinophilia), is a 594 amino acid nuclear protein that is a component of the CPSF complex. Within the complex, FIP1L1 contributes to the poly(A) recognition and stimulates poly(A) addition. Fusion of the genes encoding FIP1L1 and PDGFRA due to an interstitial deletion on chromosome 4q12 is the cause of hypereosinophilia syndrome, a rare blood disorder characterized by continuous overproduction of eosinophils in the bone marrow that leads to tissue infiltration and organ damage. There are three isoforms of FIP1L1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FIP1L1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR