Anti-FLJ11184/C4orf43 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-13180R-A680
BOSSBS-13180R-A680EA 890 EUR
BOSSBS-13180R-A680
Anti-FLJ11184/C4orf43 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Representing approximately 6% of the human genome, chromosome 4 contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease. Chromosome 4 reportedly contains the largest gene deserts (regions of the genome with no protein encoding genes) and has one of the two lowest recombination frequencies of the human chromosomes. The FLJ11184 gene product has been provisionally designated FLJ11184 pending further characterization.
Type: Primary
Antigen: FLJ11184/C4orf43
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-203/203
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen