Anti-FMO3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-13186R-A750
BOSSBS-13186R-A750EA 890 EUR
BOSSBS-13186R-A750
Anti-FMO3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The Flavin containing monooxygenase family consists of five gene products, FMO1-5, that are major enzymatic oxidants involved in the metabolism of various therapeutics. Located in the liver, FMO3 is a hepatic microsomal enzyme that oxygenates soft nucleophiles such as secondary and tertiary amines. Through its N-oxygenase capabilities, FMO3 acts on a variety of xenobiotics to catalyze oxidative digestion. Defects in the FMO3 gene are the primary cause of trimethylaminuria (TMAuria), an inborn error of metabolism associated with a fishy body odor emitting from sweat, urine and breath. Genetic mutations in FMO3 lead to the N-oxidation of amino-trimethylamine derived from food products, thus producing the malodor associated with TMAuria.
Type: Primary
Antigen: FMO3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 111-210/532
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen