Anti-G6PC3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-13253R-A647
BOSSBS-13253R-A647EA 893 EUR
BOSSBS-13253R-A647
Anti-G6PC3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Glucose-6-phosphatase (G6Pase), is a multicomponent enzyme system that hydrolyzes glucose-6-phosphate (G6P) in the final step of gluconeogenesis and gluconeolysis. G6Pase localizes to the endoplasmic reticulum, and while liver, kidney, and intestine are the only tissues that express the first identified isoform, G6Pase-Alpha, a second form, designated G6Pase-Beta, contributes to blood glucose homeostasis in a wider range of tissues. G6Pase-Beta, also known as SCN4, UGRP or G6PC3 (glucose 6 phosphatase, catalytic, 3), is a 346 amino acid endoplasmic reticulum multi-pass membrane protein that is involved in carbohydrate biosynthesis and the gluconeogenesis pathway. Inhibited by vanadate, G6Pase-Beta hydrolyzes GP6 to glucose in the endoplasmic reticulum. Due to its necessary involvement in normal glucose metabolism, G6Pase-Beta may play an integral role in diabetes and glycogen storage diseases (GSDs).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G6PC3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR