Anti-GARNL3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-13285R-A647
BOSSBS-13285R-A647EA 893 EUR
BOSSBS-13285R-A647
Anti-GARNL3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
GARNL3

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GARNL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR