Anti-GATM Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-13296R-A555
BOSSBS-13296R-A555EA 893 EUR
BOSSBS-13296R-A555
Anti-GATM Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Catalyzes the biosynthesis of guanidinoacetate, the immediate precursor of creatine. Creatine plays a vital role in energy metabolism in muscle tissues. May play a role in embryonic and central nervous system development. May be involved in the response to heart failure by elevating local creatine synthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GATM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR