Anti-GATM Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13296R-A680
BOSSBS-13296R-A680EA 890 EUR
BOSSBS-13296R-A680
Anti-GATM Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Catalyzes the biosynthesis of guanidinoacetate, the immediate precursor of creatine. Creatine plays a vital role in energy metabolism in muscle tissues. May play a role in embryonic and central nervous system development. May be involved in the response to heart failure by elevating local creatine synthesis.

Type: Primary
Antigen: GATM
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 141-240/423
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR