Anti-GATM Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-13296R-CY7
BOSSBS-13296R-CY7EA 900 EUR
BOSSBS-13296R-CY7
Anti-GATM Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Catalyzes the biosynthesis of guanidinoacetate, the immediate precursor of creatine. Creatine plays a vital role in energy metabolism in muscle tissues. May play a role in embryonic and central nervous system development. May be involved in the response to heart failure by elevating local creatine synthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GATM
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR