Anti-GCHFR Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13313R
BOSSBS-13313REA 725 EUR
BOSSBS-13313R
Anti-GCHFR Rabbit Polyclonal Antibody
Antikörper
GTP cyclohydrolase I feedback regulatory protein (GFRP) is encoded by the gene GCHFR. GFRP mediates feedback inhibition of GTP cyclohydrolase I activity by tetrahydrobiopterin. GFRP also acts as a mediator for the stimulatory effect of phenylalanine on enzyme activity. L-phenylalanine reverses this inhibition. The molecular weight of GFRP is approximately 9.5kDa although cross-linking experiments have shown that GFRP is usually found as a homodimer or as a pentamer. 15q15.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GCHFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR