Anti-GCNT3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-13317R-FITC
BOSSBS-13317R-FITCEA 900 EUR
BOSSBS-13317R-FITC
Anti-GCNT3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
2010013H22Rik; 2210021I22Rik; 2210401J11Rik; 3-galactosyl-O-glycosyl-glycoprotein beta-1; 6-N-acetylglucosaminyltransferase 3; 6-N-acetylglucosaminyltransferase; Beta 1 3 galactosyl O glycosyl glycoprotein beta 1 6 N acetylglucosaminyltransferase 3; Beta-1; Beta1 6 N acetylglucosaminyltransferase 3; beta1 6 N acetylglucosaminyltransferase; C2/4GnT; C24GNT; C2GnT M; C2GnT mucin type; C2GnT-M; C2GnT-mucin type; C2GnT2; C2GNTM; Core 2 beta 1 6 N acetylglucosaminyltransferase II; Core 2/core 4 beta 1 6 N acetylglucosaminyltransferase; Core 2/core 4 beta-1; dI/C2/C4GnT; EC 2.4.1.102; EC 2.4.1.150; GCNT3; GCNT3_HUMAN; Glucosaminyl (N acetyl) transferase 3; Glucosaminyl (N acetyl) transferase 3 mucin type; GnT M; GNTM; hC2GnT M; hC2GnT-M; Mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase. OTTHUMP00000163601.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GCNT3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR