Anti-GFPT2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-13342R
BOSSBS-13342REA 1090 EUR
BOSSBS-13342R
Anti-GFPT2 Rabbit Polyclonal Antibody
Antikörper
GFPT2 (glutamine-fructose-6-phosphate transaminase 2), also known as D-fructose-6-phosphate amidotransferase 2 or hexosephosphate aminotransferase 2, is a 682 amino acid protein and isoenzyme of GFAT1, the first and rate-limiting enzyme for the entry of glucose into the hexosamine biosynthetic pathway (HBP), which is a relatively minor branch of glycolysis. Expressed in spinal cord, heart and placenta, GFAT2 regulates glucose entry into the HBP and likely controls the availability of precursors for N- and O-linked protein glycosylation. Containing one glutamine amidotransferase type-2 domain and two SIS domains. GFAT2 is encoded by a gene that maps to human chromosome 5q35.3. GFAT2 gene variants have been linked to type 2 diabetes, diabetic nephropathy, and increased GFPT2 mRNA levels.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GFPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR