Anti-GGCT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-13346R-A488
BOSSBS-13346R-A488EA 893 EUR
BOSSBS-13346R-A488
Anti-GGCT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Catalyzes the formation of 5-oxoproline from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis. Induces release of cytochrome c from mitochondria with resultant induction of apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GGCT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat