Anti-GGCT Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13346R-A680
BOSSBS-13346R-A680EA 890 EUR
BOSSBS-13346R-A680
Anti-GGCT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Catalyzes the formation of 5-oxoproline from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis. Induces release of cytochrome c from mitochondria with resultant induction of apoptosis.

Type: Primary
Antigen: GGCT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/188
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR