Anti-GGCT Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13346R-A750
BOSSBS-13346R-A750EA 890 EUR
BOSSBS-13346R-A750
Anti-GGCT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Catalyzes the formation of 5-oxoproline from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis. Induces release of cytochrome c from mitochondria with resultant induction of apoptosis.

Type: Primary
Antigen: GGCT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/188
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

TECHNISCHE DATEN

Antigen-Name GGCT
Environmentally Preferable
Antigen-Synonyme CRF21|C7orf24|Gamma-glutamylcyclotransferase|GGCT_HUMAN|Ggct|Cytochrome c-releasing factor 21
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 750
Reaktivität
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Modification Unmodified
Epitop 51-150/188
Immunogen KLH conjugated synthetic peptide derived from human GGCT
Form Liquid
Molecular weight 21 kDa
Reinigung Purified by Protein A
Konzentration 1 µg/µl
Lagerungspuffer Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Lagertemperatur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transporttemperatur 4 °C
Validierte Anwendungen ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR