Anti-GGCT Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-13346R-CY3
BOSSBS-13346R-CY3EA 900 EUR
BOSSBS-13346R-CY3
Anti-GGCT Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Catalyzes the formation of 5-oxoproline from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis. Induces release of cytochrome c from mitochondria with resultant induction of apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GGCT
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR