Anti-GGT5 heavy chain Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13349R-A750
BOSSBS-13349R-A750EA 890 EUR
BOSSBS-13349R-A750
Anti-GGT5 heavy chain Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
g-glutamyltranspeptidase (GGT) acts as a glutathionase and catalyzes the transfer of the glutamyl moiety of Glutathione to a variety of amino acids and dipeptide acceptors. This enzyme is located on the outer surface of the cell membrane and is widely distributed in mammalian tissues involved in absorption and secretion. In humans, hepatic GGT activity is elevated in some liver diseases. GGT1 is released into the bloodstream after liver damage and an elevated level of the enzyme may be a useful early sign of hepatocellular carcinoma. GGT5 converts Leukotriene C4 to Leukotriene D4; it does not, however, convert synthetic substrates that are commonly used to assay GGT. In human serum and tissues there is a marked heterogeneity in GGT, but this heterogeneity can be attributed to different glycosylation of the same peptide rather than to the products of different genes.

Type: Primary
Antigen: GGT5 heavy chain
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/568
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR