Anti-GGTL1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-13352R-A350
BOSSBS-13352R-A350EA 893 EUR
BOSSBS-13352R-A350
Anti-GGTL1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
This gene encodes a member of the gamma-glutamyl transpeptidase (GGT) family, which are important in the metabolism of glutathione. The most ubiquitously expressed human GGT gene, GGT1, encodes a single transmembrane polypeptide that is post-translationally processed to form a heavy and a light chain. In contrast, the product of this gene only contains homology to the light chain region, and lacks a transmembrane domain. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GGTL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat