Anti-GLI4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-13373R-A647
BOSSBS-13373R-A647EA 893 EUR
BOSSBS-13373R-A647
Anti-GLI4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. GLI-4, also known as HKR4, is a 376 amino acid protein that localizes to the nucleus and contains seven C2H2-type zinc fingers. Belonging to the krueppel C2H2-type zinc-finger protein family, GLI-4 may function as a transcriptional regulator,More Product Information