Anti-GOX Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-13387R-A488
BOSSBS-13387R-A488EA 893 EUR
BOSSBS-13387R-A488
Anti-GOX Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Glucose Oxidase is a dimeric enzyme that binds to β-D-glucose and aids in its oxidation into D-glucono-1,5-lactone which then hydrolyzes to gluconic acid. Flavin adenine dinucleotide (FAD) is a cofactor to Glucose Oxidase that acts as the initial electron acceptor and is required for this oxidation to occur. Glucose Oxidase is a natural preservative found in honey, where it reduces atmospheric oxygen into hydrogen peroxide which acts as an antibacterial barrier. Glucose Oxidase is also commonly used in biosensors in which it conveys levels of glucose by keeping track of the number of electrons passed through the enzyme. In this application, Glucose Oxidase is connected to an electrode and the resulting charge is measured.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glucose Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR