Anti-EPRS Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-13393R-A555
BOSSBS-13393R-A555EA 893 EUR
BOSSBS-13393R-A555
Anti-EPRS Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. Aminoacyl-tRNA synthetases function to catalyze the aminoacylation of tRNAs by their corresponding amino acids, thus linking amino acids with tRNA-contained nucleotide triplets. ProRS (Prolyl-tRNA synthetase), also known as EPRS, EARS, PARS, QARS, QPRS, PIG32 or GLUPRORS, is a 1,512 amino acid protein that contains three WHEP-TRS domains and belongs to both the class-I and class-II aminoacyl-tRNA synthetase family. Functioning as a component of the multisynthase complex, ProRS uses ATP to catalyze the conversion of L-glutamate and tRNA(Glu) to L-glutamyl-tRNA(Glu), as well as the conversion of L-proline and tRNA(Pro) to L-prolyl-tRNA(Pro).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPRS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR