Anti-GSTZ1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-13442R-A647
BOSSBS-13442R-A647EA 893 EUR
BOSSBS-13442R-A647
Anti-GSTZ1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
GSTZ1 is a member of the glutathione S transferase (GSTs) super family, encoding multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Several transcript variants of this gene encode multiple protein isoforms. GSTZ1 shows minimal glutathione-conjugating activity with ethacrynic acid and 7-chloro-4-nitrobenz-2-oxa-1,3-diazole and maleylacetoacetate isomerase activity. It has low glutathione peroxidase activity with T butyl and cumene hydroperoxides and is able to catalyze the glutathione dependent oxygenation of dichloroacetic acid to glyoxylic acid. Highest expression in liver followed by kidney, skeletal muscle and brain. Also expressed in melanocytes, synovium, placenta, breast and fetal liver and heart.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glutathione Transferase zeta 1/MAAI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR