Anti-GMDS Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-13453R-A680
BOSSBS-13453R-A680EA 890 EUR
BOSSBS-13453R-A680
Anti-GMDS Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Catalyzes the conversion of GDP-D-mannose to GDP-4-dehydro-6-deoxy-D-mannose.
Type: Primary
Antigen: GMDS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/372
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen