Anti-GMDS Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13453R-A750
BOSSBS-13453R-A750EA 890 EUR
BOSSBS-13453R-A750
Anti-GMDS Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Catalyzes the conversion of GDP-D-mannose to GDP-4-dehydro-6-deoxy-D-mannose.

Type: Primary
Antigen: GMDS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/372
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR