Anti-GMP Synthase Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13459R-A680
BOSSBS-13459R-A680EA 890 EUR
BOSSBS-13459R-A680
Anti-GMP Synthase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
In the de novo synthesis of purine nucleotides, IMP is the branch point metabolite at which point the pathway diverges to the synthesis of either guanine or adenine nucleotides. In the guanine nucleotide pathway, there are 2 enzymes involved in converting IMP to GMP, namely IMP dehydrogenase (IMPD1), which catalyzes the oxidation of IMP to XMP, and GMP synthetase, which catalyzes the amination of XMP to GMP.

Type: Primary
Antigen: GMP Synthase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/693
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR