Anti-GMPPA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-13460R-A555
BOSSBS-13460R-A555EA 893 EUR
BOSSBS-13460R-A555
Anti-GMPPA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
GMPPA is a GDP-mannose pyrophosphorylase which catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose, in the production of N-linked oligosaccharides.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GMPPA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat