Anti-GNGT1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-13470R-CY5
BOSSBS-13470R-CY5EA 900 EUR
BOSSBS-13470R-CY5
Anti-GNGT1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Heterotrimeric guanine nucleotide-binding proteins (G proteins) transduce extracellular signals received by transmembrane receptors to effector proteins. Transducin is a guanine nucleotide-binding protein found specifically in rod outer segments, where it mediates activation by rhodopsin of a cyclic GTP-specific (guanosine monophosphate) phosphodiesterase. Transducin is also referred to as GMPase. GNGT1 encodes the gamma subunit of transducin (Hurley et al., 1984 [PubMed 6438626]; Scherer et al., 1996 [PubMed 8661128]).[supplied by OMIM, Mar 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNGT1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR