Anti-Glucosamine 6 sulfatase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13479R-A750
BOSSBS-13479R-A750EA 890 EUR
BOSSBS-13479R-A750
Anti-Glucosamine 6 sulfatase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
GNS is a 552 amino acid lysosomal enzyme that hydrolyzes the 6-sulfate groups of the N-acetyl-D-glucosamine 6-sulfate units of keratan sulfate and heparan sulfate. A member of the sulfatase family, GNS assists in the catabolism of heparin, and binds calcium as a cofactor. GNS deficiency results in an autosomal recessive lysosomal storage disorder known as mucopolysaccharidosis type IIID (Sanfilippo D syndrome), which is characterized by mild somatic disease and severe degeneration of the central nervous system. Subject to post-translational internal peptidase cleavage, GNS is encoded by a gene mapping to human chromosome 12q14.2 and mouse chromosome 10 D2.

Type: Primary
Antigen: Glucosamine 6 sulfatase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/552
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR